ENA410543204 Enamine

3-[(4-chlorophenyl)sulfanyl]-4-nitrobenzoic acid

  • MDL number MFCD22483356
  • Cas Number : 1553287-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139732
Availability details for
Quantity
  • Availability
  • Add to Cart