ENA410543205 Enamine

6-chloro-5-methoxypyridine-3-sulfonyl chloride

  • MDL number MFCD17014999
  • Cas Number : 75720-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139733
Availability details for
Quantity
  • Availability
  • Add to Cart