ENA410543207 Enamine

3-(2-methylpropyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD12137243
  • Cas Number : 1153977-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139735

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139735
Availability details for
Quantity
  • Availability
  • Add to Cart