ENA410543212 Enamine

methyl 2-(tert-butoxy)acetate

  • MDL number MFCD11196078
  • Cas Number : 137676-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139752

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139752
Availability details for
Quantity
  • Availability
  • Add to Cart