ENA410543227 Enamine

2-(3-bromo-5-fluorophenyl)pyrrolidine

  • MDL number MFCD11619365
  • Cas Number : 1094510-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139852
Availability details for
Quantity
  • Availability
  • Add to Cart