ENA410543236 Enamine

2-(2,3-dihydro-1H-inden-1-yl)guanidine hydrochloride

  • MDL number MFCD26408032
  • Cas Number : 1607290-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139872
Availability details for
Quantity
  • Availability
  • Add to Cart