ENA410543238 Enamine

ethyl 2-bromo-3-chloro-5-methylbenzoate

  • MDL number MFCD24108610
  • Cas Number : 1508456-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139875
Availability details for
Quantity
  • Availability
  • Add to Cart