ENA410543241 Enamine

7-hydroxynaphthalene-1-carbaldehyde

  • MDL number MFCD17012512
  • Cas Number : 144876-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139899

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139899
Availability details for
Quantity
  • Availability
  • Add to Cart