ENA410543244 Enamine

1-(3,5,6-trimethylpyrazin-2-yl)methanamine

  • MDL number MFCD17018339
  • Cas Number : 866751-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139920
Availability details for
Quantity
  • Availability
  • Add to Cart