ENA410543251 Enamine

3-(chloromethyl)-1-(4-chlorophenyl)-1H-pyrazole

  • MDL number MFCD21304457
  • Cas Number : 1481659-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139958
Availability details for
Quantity
  • Availability
  • Add to Cart