ENA410543252 Enamine

6-bromo-N-methylpyridin-2-amine

  • MDL number MFCD09261221
  • Cas Number : 89026-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139967

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139967
Availability details for
Quantity
  • Availability
  • Add to Cart