ENA410543253 Enamine

5,7-dichloro-3-(propan-2-yl)pyrazolo[1,5-a]pyrimidine

  • MDL number MFCD12923044
  • Cas Number : 771510-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139968
Availability details for
Quantity
  • Availability
  • Add to Cart