ENA410543260 Enamine

1-[(2-fluorophenyl)methyl]-4-methyl-1H-pyrazol-5-amine

  • MDL number MFCD10691733
  • Cas Number : 1152525-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139977
Availability details for
Quantity
  • Availability
  • Add to Cart