ENA410543265 Enamine

1-methyl-2-(oxane-4-carbonyl)-1H-imidazole

  • MDL number MFCD21188318
  • Cas Number : 1492724-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139987
Availability details for
Quantity
  • Availability
  • Add to Cart