ENA410543270 Enamine

sodium 2-[4-(naphthalen-1-yl)-1,3-thiazol-2-yl]acetate

  • MDL number MFCD26408045
  • Cas Number : 1803595-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140015
Availability details for
Quantity
  • Availability
  • Add to Cart