ENA410543274 Enamine

3,3,5,5-tetramethylpiperidine hydrochloride

  • MDL number MFCD26408047
  • Cas Number : 792915-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 251...253
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140086

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140086
Availability details for
Quantity
  • Availability
  • Add to Cart