ENA410543280 Enamine

4-(1H-pyrrol-1-yl)benzene-1-sulfonamide

  • MDL number MFCD01762129
  • Cas Number : 5438-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237...239
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140096

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140096
Availability details for
Quantity
  • Availability
  • Add to Cart