ENA410543289 Enamine

4-phenylpiperidin-4-ol hydrochloride

  • MDL number MFCD21605333
  • Cas Number : 5004-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140123
Availability details for
Quantity
  • Availability
  • Add to Cart