ENA410543290 Enamine

[2-methyl-4-(trifluoromethyl)-1,3-thiazol-5-yl]methanol hydrochloride

  • MDL number MFCD26408052
  • Cas Number : 1803584-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140125

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140125
Availability details for
Quantity
  • Availability
  • Add to Cart