ENA410543294 Enamine

3-{[(2-methylphenyl)methanesulfinyl]methyl}furan-2-carboxylic acid

  • MDL number MFCD17386446
  • Cas Number : 1274622-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140139
Availability details for
Quantity
  • Availability
  • Add to Cart