ENA410543310 Enamine

1-(4-trifluoromethanesulfonylphenyl)piperazine hydrochloride

  • MDL number MFCD26408066
  • Cas Number : 1803598-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140244
Availability details for
Quantity
  • Availability
  • Add to Cart