ENA410543311 Enamine

4-butylmorpholine-3-carboxylic acid hydrochloride

  • MDL number MFCD26408068
  • Cas Number : 1607020-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140329
Availability details for
Quantity
  • Availability
  • Add to Cart