ENA410543315 Enamine

[2-methyl-4-(trifluoromethyl)-1,3-thiazol-5-yl]methanesulfonyl chloride

  • Molecular Weight : 279.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-4-(trifluoromethyl)thiazol-5-yl)methanesulfonyl chloride
Inchi Code 1S/C6H5ClF3NO2S2/c1-3-11-5(6(8,9)10)4(14-3)2-15(7,12)13/h2H2,1H3
Inchi Key NLACIVCDBBHNKM-UHFFFAOYSA-N

Description

Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-140929
hstatements H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart