ENA410543320 Enamine

1-methyl-3-(propan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • MDL number MFCD22718116
  • Cas Number : 2019997-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140206
Availability details for
Quantity
  • Availability
  • Add to Cart