ENA410543321 Enamine

2-fluoro-4-(methoxycarbonyl)benzoic acid

  • MDL number MFCD20640446
  • Cas Number : 314241-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140207

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-140207
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart