ENA410543330 Enamine

2-[(pyridin-3-yl)methyl]aniline dihydrochloride

  • MDL number MFCD26408061
  • Cas Number : 1803610-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140231
Availability details for
Quantity
  • Availability
  • Add to Cart