ENA410543331 Enamine

[2-(phenoxymethyl)-1,3-thiazol-4-yl]methanamine hydrochloride

  • MDL number MFCD26408062
  • Cas Number : 1797023-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140232
Availability details for
Quantity
  • Availability
  • Add to Cart