ENA410543334 Enamine

5-(2,4-dimethylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD00629032
  • Cas Number : 1182997-40-5
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-140180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140180
Availability details for
Quantity
  • Availability
  • Add to Cart