ENA410543335 Enamine

5-amino-N-cyclopropyl-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide

  • MDL number MFCD21038619
  • Cas Number : 1451993-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140181
Availability details for
Quantity
  • Availability
  • Add to Cart