ENA410543336 Enamine

2-(cyclopropylmethyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD16698620
  • Cas Number : 1249719-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140298
Availability details for
Quantity
  • Availability
  • Add to Cart