ENA410543337 Enamine

2-[3-(trifluoromethyl)phenyl]piperidine hydrochloride

  • MDL number MFCD02178967
  • Cas Number : 1184989-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140300
Availability details for
Quantity
  • Availability
  • Add to Cart