ENA410543339 Enamine

N-(3-aminopyridin-4-yl)-4-ethoxybenzamide

  • MDL number MFCD18059222
  • Cas Number : 1281215-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140302

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140302
Availability details for
Quantity
  • Availability
  • Add to Cart