ENA410543340 Enamine

N-[2-(3-aminobenzenesulfonamido)ethyl]acetamide

  • MDL number MFCD12433136
  • Cas Number : 1157001-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140304
Availability details for
Quantity
  • Availability
  • Add to Cart