ENA410543343 Enamine

methyl 5-(3-chlorophenyl)-1H-pyrrole-2-carboxylate

  • MDL number MFCD26408056
  • Cas Number : 1797395-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140182
Availability details for
Quantity
  • Availability
  • Add to Cart