ENA410543352 Enamine

3-(3-chlorophenyl)-4,5-dihydro-1,2-oxazol-5-one

  • MDL number MFCD11845106
  • Cas Number : 51725-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140185

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140185
Availability details for
Quantity
  • Availability
  • Add to Cart