ENA410543353 Enamine

3-methoxy-2-methylpropane-1-sulfonyl chloride

  • MDL number MFCD19681320
  • Cas Number : 1340102-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140186
Availability details for
Quantity
  • Availability
  • Add to Cart