ENA410543363 Enamine

N-methyl-4-oxo-3,4-dihydrophthalazine-1-carboxamide

  • MDL number MFCD07355819
  • Cas Number : 103264-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15601
Availability details for
Quantity
  • Availability
  • Add to Cart