ENA410543499 Enamine

(2E)-2-hydrazinylidene-1,2-diphenylethan-1-one

  • MDL number MFCD00007625
  • Cas Number : 5344-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17365

Safety Information

msds https://www.enaminestore.com/catalog/EN300-17365
Availability details for
Quantity
  • Availability
  • Add to Cart