ENA410543821 Enamine

2-(carboxymethyl)-1,4-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD00010742
  • Cas Number : 33369-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-20522

Safety Information

safety_statement 26-37/39
msds https://www.enaminestore.com/catalog/EN300-20522
Availability details for
Quantity
  • Availability
  • Add to Cart