ENA410543881 Enamine

cyclopropyldiphenylmethanol

  • MDL number MFCD00001298
  • Cas Number : 5785-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...87
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21005

Safety Information

msds https://www.enaminestore.com/catalog/EN300-21005
Availability details for
Quantity
  • Availability
  • Add to Cart