ENA410543995 Enamine

5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazolidin-4-yl}pentanoic acid

  • MDL number MFCD00128981
  • Cas Number : 57378-70-8
  • Molecular Weight : 244.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224-226
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23975

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-23975
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart