ENA410544024 Enamine

5-amino-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD00022384
  • Cas Number : 28491-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33948

Safety Information

signalword Warning
safety_statement 26-37/39
msds https://www.enaminestore.com/catalog/EN300-33948
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart