ENA410544053 Enamine

(4-ethoxyphenyl)methanol

  • MDL number MFCD00004655
  • Cas Number : 6214-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32...34
Purity 95.00
Flash Point 113
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49131

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-49131
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart