ENA410544064 Enamine

4-(difluoromethoxy)-3-(trifluoromethyl)aniline

  • MDL number MFCD14650280
  • Cas Number : 71791-37-2
  • Molecular Weight : 227.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52691
Availability details for
Quantity
  • Availability
  • Add to Cart