ENA410544070 Enamine

3-(2-nitroethenyl)thiophene

  • Molecular Weight : 155.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55573
Availability details for
Quantity
  • Availability
  • Add to Cart