ENA410544071 Enamine

2-[(4-aminophenyl)(methyl)amino]ethan-1-ol

  • MDL number MFCD11184967
  • Cas Number : 17625-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55760
Availability details for
Quantity
  • Availability
  • Add to Cart