ENA410544072 Enamine

2,2,2-trifluoroethyl N-(6-chloropyridin-3-yl)carbamate

  • MDL number MFCD26147049
  • Cas Number : 1551362-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56738
Availability details for
Quantity
  • Availability
  • Add to Cart