ENA410544107 Enamine

N-(6-chloropyrazin-2-yl)acetamide

  • MDL number MFCD01201713
  • Cas Number : 132453-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140190
Availability details for
Quantity
  • Availability
  • Add to Cart