ENA410544121 Enamine

tert-butyl N-(2-hydroxypropyl)carbamate

  • MDL number MFCD02093569
  • Cas Number : 95656-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95742
Availability details for
Quantity
  • Availability
  • Add to Cart