ENA410544128 Enamine

3-[3-(trifluoromethyl)phenyl]propanoic acid

  • MDL number MFCD00090487
  • Cas Number : 585-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98646

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-98646
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart